Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548929

Catalog No. PA548929

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Does not catalyze the synthesis of 1-aminocyclopropane- 1-carboxylate but is capable of catalyzing the deamination of L- vinylglycine.
Specifications

Specifications

ACCS
Polyclonal
PBS with 0.09% sodium azide
Q96QU6
1-aminocyclopropane-1-carboxylate synthase-like protein 1, ACC synthase-like protein 1, ACCS, PHACS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84680
Western Blot
Unconjugated
ACCS
Liquid
ACCS
KLH conjugated synthetic peptide between 224-250 amino acids from the Central region of human ACCS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit