Learn More
Bioscience accu-Test™ Vials for COD Testing
For low-, standard- and high-range concentrations
Manufacturer:  Bioscience 185 240
 View more versions of this product
Catalog No. 01-215-29
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Includes: 16mm vial with 1.5mL digestion solution, 3.5mL catalyst solution, and screw cap
Description
Description
  • Convenient to use
  • Give excellent analytical results
  • Compatible with all standard COD reactors and spectrophotometers
  • For samples with low chloride and ammonia levels, mercury-free reagents are available for nonreporting purposes
Order Info
Supplied in modular packs that are easy to stack
Certifications
U.S. EPA approved (for standard and low ranges) micro Chemical Oxygen Demand test
Specifications
Specifications
EPA, OSHA
COD testing for 5 to 150mg/L standard range concentration
16mm vial with 1.5mL digestion solution, 3.5mL catalyst solution, and screw cap
75% Sulfuric Acid, 0.20% Potassium Dichromate, 0.40% Silver Sulfate, 0.002%Sulfamic Acid
Standard COD reactor and spectrophotometer