Learn More
Thermo Scientific™ Accucore™ HILIC LC Columns Available on GSA/VA Contract for Federal Government customers only.
Offer enhanced retention of polar and hydrophilic analytes with Thermo Scientific™ Accucore™ HILIC LC Columns.
Manufacturer:  Thermo Scientific™ 17526014005
 View more versions of this product
Catalog No. 17-526-014005
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Offer enhanced retention of polar and hydrophilic analytes with Thermo Scientific™ Accucore™ HILIC LC Columns. Based on Core Enhanced Technology™, Accucore columns provide fast, high-resolution separations without the elevated backpressures required by sub-2µm particles.
  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Enhanced Retention of Polar and Hydrophilic Analytes
  • Alternative Selectivity to C18 without Ion-pair or Derivatisation
  • Improved Sensitivity for MS detection
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Description & Specifications
Specifications
Guard Cartridge, 4 PK
4mm
1000bar
Spherical Solid Core
2 to 8
130m²/g
L3
HILIC
10mm
Silica, Spherical Solid Core Ultrapure
2.6μm
80Å
70°C
Silica