Learn More
Thermo Scientific™ Accucore™ Urea HILIC LC Columns Available on GSA/VA Contract for Federal Government customers only.
Offer unique HILIC selectivity and low ion exchange activity with Thermo Scientific™ Accucore™ Urea HILIC LC Columns.
 View more versions of this product
Catalog No. 03-452-128
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Based on Core Enhanced Technology™, Thermo Scientific™ Accucore™ Urea HILIC LC Columns provide fast, high-resolution separations-without the elevated backpressures required by sub-2µm particles.
  • Rugged and reproducible 2.6µm solid core particles
  • Fast separations with superb resolution
  • Low backpressures
  • Bonded hydrophillic stationary phase
  • Unique selectivity compared to other HILIC phases
  • Low ion exchange activity
Description & Specifications
Specifications
Analytical Column
2.1mm
1000bar
Spherical Solid-core
2 to 8
130m²/g
N/A
HILIC
150mm
Silica, Spherical Solid Core Ultrapure
2.6μm
80Å
70°C
Urea-HILIC