Learn More
Thermo Scientific™ Accucore™ Wide Selectivity Kits Available on GSA/VA Contract for Federal Government customers only.
Verify optimum selectivity of Thermo Scientific™ Accucore™ HPLC columns over a wide range.
Manufacturer:  Thermo Scientific™ 17X260521303VB
 View more versions of this product
Catalog No. 03-051-907
Add to cart
Description
Description
Use the Thermo Scientific™ Accucore™ Wide Selectivity Kits to verify which Accucore column provides optimum selectivity for your application. Each kit includes 1 Accucore C18, 1 Accucore PhenylHexyl, and 1 Accucore PHP HPLC column. Accucore columns are based on advanced Core Enhanced Technology™ to provide fast, high-resolution separations with excellent resolution and sensitivity. Although sub 2um-particle columns typically produce high background pressures, these Accucore columns produce lower backpressures; UHPLC systems are therefore not required for use with these columns.
  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Kit contains 1 Accucore C18 HPLC Column, 1 Accucore PhenylHexyl Column and 1 Accucore PHP Column
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2.1mm
HPLC
50mm
2.6μm
130m²/g
Yes
1x Accucore C18 column
1x Accucore PhenylHexyl column
1x Accucore PHP column
Spherical Solid Core
80Å
Accucore™ C18, PFP, Phenyl/Hexyl