Learn More
Fisherbrand™ AccuDuster™ AccuDuster III Air Duster
The AccuDuster III is compressed gas for precision cleaning of data processing and electronic equipment. This duster featuring a trigger-operated valve and one-inch extension wand can be used for speed drying of wet surfaces and for removal of dust, lint and oxide particles from electronic equipment, optical surfaces, precision instruments and PC boards.
Catalog No. 18-431
Add to cart
Description
Description
  • Removes dust and other contaminants not accessible by other cleaning agents
  • Trigger-operated valve delivers fingertip control
  • Includes extension wand for hard to reach places
  • 71 psi pressure at room temperature
  • Precision filtered to less than 0.2 microns
  • Contains HFCs
  • Nonflammable
Specifications
Specifications
10 oz.
10 oz., 295.73mL
Removes dust and other contaminants not accessible by other cleaning agents
Air duster
295.73mL
No
71psi