Learn More
Getinge USA ACCUFAST TEST PK 25CTRLS 25/CS  DFS Item
Manufacturer:  Getinge USA 61301606639
GETINGE ASSURE ACCUFAST. The first and only early release test system offering true biological confirmation, Getinge Assure AccuFast combines a rapid biological indicator (BI) with a state-of-the-art chemical integrator (CI) for sterility assurance that's fast, accurate and verifiable. Because timing is critical in BI monitoring, AccuFast can provide failed cycle notification in as little as 3-5 hours. Ideal for monitoring all types of steam sterilization cycles. Gain the power to confidently release items for use with patients, eliminate any guesswork, improve patient safety, and lower costs.
Catalog No. NC0800091