Learn More
Accuform Signs 2.5 - 5 in. Lockout Gate Valves
Manufacturer:  ACCUFORM SIGNS KDD402YL
 View more versions of this product
Catalog No. 19-090-401
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yellow
2.5 to 5 in.
-50° to 360°F
Polypropylene
To enclose and completely surround the valve handles
6.35 to 13cm
-46° to 182°C
Lockout Gate Valve