Accuform Signs Blank NFPA Placards

Manufacturer: ACCUFORM SIGNS MBLEV15

 View more versions of this product

Catalog No. 19-804-655

  Check Availability
Add to Cart

    Specifications

    Includes Placard only, hazard panels are 2mil clear Polyester
    Height (English) 15 in.
    Material Vinyl
    Message NFPA
    Quantity Each
    Color Red, Blue, Yellow on White,Red, Blue, Yellow on White
    Type NFPA placard
    Width (English) 15 in.
    For Use With (Application) Designed for on-the-spot classification of potentially hazardous chemicals

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit