Learn More
Accuform Signs Blank Tags
Use Blank Tags to clearly identify potential hazards in and around your workplace.
$33.95 - $75.30
Specifications
Diameter (English) Grommet | 0.37 in. |
---|---|
For Use With (Application) | For indoor and outdoor conditions |
Length (Metric) | 14.6 cm |
Length (English) | 5.75 in. |
Width (English) | 3.25 in. |
Catalog Number | Mfr. No. | Color | Material | Message | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Material | Message | Price | Quantity & Availability | |||
17-998-152
|
Accuform Signs
MDT260CTP |
Black, Red, White | Pulp-Free Cardstock | DANGER |
Pack of 25 for $33.95
|
|
|||
17-998-153
|
Accuform Signs
MDT260PTP |
Black, Red, White | RP Plastic | DANGER |
Pack of 25 for $75.30
|
|
|||
17-998-156
|
Accuform Signs
MNT101CTP |
Blue, White | Pulp-Free Cardstock | Notice |
Pack of 25 for $33.95
|
|
|||
17-998-154
|
Accuform Signs
MWGT205CTP |
Black, Orange | Pulp-Free Cardstock | WARNING |
Pack of 25 for $33.95
|
|
|||
17-998-155
|
Accuform Signs
MWGT205PTP |
Black, Orange | RP Plastic | WARNING |
Pack of 25 for $75.30
|
|
|||
17-998-157
|
Accuform Signs
MNT101PTP |
Blue, White | RP Plastic | Notice |
Pack of 25 for $75.30
|
|
|||
Specifications
0.37 in. | |
14.6 cm | |
3.25 in. | |
5.75 x 3.25 in. (14.6 x 8.25 cm) |
For indoor and outdoor conditions | |
5.75 in. | |
8.25 cm | |
0.93 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.