Accuform Signs Caution Watch Your Step Sign

Colorful, compact safety signs


 View all options for this product

Catalog No. 19-804-704


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Certifications/Compliance ANSI Z535.2-22, 29 CFR 191.145
    Color Black on Yellow
    Dimensions (L x W) 10 x 14 in. (25.4 x 35.6cm)
    For Use With (Application) Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage
    Height (English) 10 in.
    Languages English
    Length (English) 10 in.
    Length (Metric) 25.4cm
    Material Plastic
    Message Watch Your Step
    Quantity Each
    Style Surface Mount
    Temperature Range (English) -90 to 168°F
    Temperature Range (Metric) -68 to 76°C
    Thickness 0.06 in.
    Type Safety sign
    Width (English) 14 in.
    Width (Metric) 35.6cm

    • Size: 25.4 x 35.6cm (10 x 14 in.)
    • Color: Yellow, Black
    • Language: English

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit