Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Chilled Water Return Pipe Markers
Identify contents and mark the directional flow for pipes used for chilled water return.
Supplier: Accuform Signs RPK201SSD
Specifications
Pipe Marker | |
Vinyl, Plastic | |
Clearly identifying the pipe contents and flow direction, make maintaining facility easier and increase employee safety by clearly marking pipes | |
1 Ea. | |
-67.77 to 75.55°C | |
2.5 to 6 in. |
Self-adhesive | |
Chilled Water Return | |
1 to 3 Years | |
-90 to 168°F | |
4 mil | |
6.35 to 15.24 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title