Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Color Blank Tags
These colorful blank tags allow you to get descriptive in explaining important facts
Supplier: Accuform Signs TBB302RDP
Specifications
Red | |
Cardstock | |
For coding, marking, quick identification | |
0.45 cm |
0.18 in. | |
Blank | |
Plain Hole |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction