Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Cylinder Status Tags

Cylinder Status Tags inform others if the contents are empty, full, or in use

Manufacturer:  ACCUFORM SIGNS MGT203CTP

 View more versions of this product

Catalog No. 17-998-162


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

White on Green
0.93cm
5.75 in.
10mil
3.25 in.
PF Cardstock
0.37 in.
For indoor and outdoor conditions
14.6cm
Yes
8.25cm
Full Cylinder Ready For Use
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit