Accuform Signs Danger: Compressed Air Sign

Manufacturer: ACCUFORM SIGNS LCHL149XVE

 View more versions of this product

Catalog No. 19-804-565

  Check Availability
Add to Cart

    Specifications

    Width (English) 5 in.
    Height (English) 3.5 in.
    Material Vinyl
    Message Danger Compressed Air
    Quantity Each
    Type Chemical and hazardous safety label
    For Use With (Application) Identify the danger of compressed air vessel and supply line pressure

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit