Accuform Signs Danger: Flammable Liquids


 View all options for this product

Catalog No. 17-998-071


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Certifications/Compliance ANSI Z535, OSHA 29 CFR 1910.145
    Color Red, Black on White
    For Use With (Application) Adheres to most surfaces indoor usage light duty application sheltered outdoor location
    Height (English) 7 in.
    Material Vinyl
    Message Danger Flammable Liquid
    Quantity Each
    Temperature Range (English) -20 to 146°F
    Temperature Range (Metric) -29 to 63°C
    Thickness 4mil
    Type Chemical hazard sign
    Width (English) 10 in.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit