Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Danger: High Voltage

Colorful, compact safety signs

Manufacturer:  Accuform Signs LELC036VSP

 View more versions of this product

Catalog No. 19-804-570


Add to cart

Description

Description

  • Legend: DANGER: HIGH VOLTAGE
  • Available in a variety of sizes and materials
  • Color: White, black, red
  • Language: English

Model LELC036VSP

  • Legend: DANGER. HIGH VOLTAGE INSIDE

Specifications

Specifications

Safety
Red, Black on White
English
5 Pack
5 in.
176°F
Danger High Voltage
-28°C
Electrical Hazard Sign
17.8cm
OSHA 29 CFR 1910.145
Non-glare
Vinyl
Provide proper protection in certain areas in order to stay safe on the job or in the for short term outdoor and self sticking applications
12.7cm
80°C
-20°F
4mil
7 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit