Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Danger: No Trespassing

Manufacturer:  ACCUFORM SIGNS MADM292VP

 View more versions of this product

Catalog No. 17-998-048


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ANSI Z535, OSHA 29 CFR 1910.145
Indoor usage medium-duty marking needs ideal for indoor and short-term, sheltered outdoor environments
Plastic
Each
-90 to 168°F
0.05 in.
10 in.
Red, Black on White
7 in.
Danger No Trespassing
Surface Mount
-67 to 76°C
Admittance sign
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit