Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Directional Flow Arrow Tape

Provides 360° visibility

Manufacturer:  ACCUFORM SIGNS RAW254RDWT

 View more versions of this product

Catalog No. 19-169-547


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Self-adhesive
ANSI
54 ft.
6mil
Tape
Directional Flow Arrow Tape
Red,White
Provide 360° visibility and identify flow direction with printed arrow tapes
16.45m
2 in.
Vinyl
Each
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit