Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Do Not Enter Tags
Prevent others from entering potentially hazardous areas with this safety tag.
Supplier: Accuform Signs MDT164CTP
Specifications
OSHA 29 CFR 1910.145, ANSI Z535 | |
PF-Cardstock | |
Danger Do Not Enter | |
25/Pk. | |
5.75 in. | |
3.25 in. |
Black, Red, White | |
Safety Tag | |
To identify clearly and alert staff to cautions | |
10 mil | |
14.6 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title