Accuform Signs Exit Sign

Manufacturer: ACCUFORM SIGNS MEXT06BVS

 View more versions of this product

Catalog No. 19-804-751

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Certifications/Compliance OSHA 29 CFR 1910.145
    Color Red, White
    Finish Non Glare
    For Use With (Application) For short term outdoor applications
    Height (English) 10 in.
    Length (English) 14 in.
    Material Vinyl
    Message Exit
    Quantity Each
    Thickness 4mil
    Type Safety sign

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit