Accuform Signs Fold-ups Signs - Multiple Messages

Identify temporary hazardous conditions or display messages quickly and easily

Manufacturer: ACCUFORM SIGNS PFR242

 View all options for this product

Catalog No. 19-120-2791

  Check Availability
Add to Cart


    Color Black, Red on White
    For Use With (Application) Free standing signs allow you to identify temporary hazardous conditions or display important messages, quickly and easily
    Height (English) 20 in.
    Material Plastic
    Message Decon Area
    Quantity Each
    Size (English) 12 x 20 in.
    Type Fold ups floor safety sign
    Width (English) 12 in.

    • Double-sided signs can be set-up and moved easily to exactly where they are needed
    • Rugged, high-impact plastic construction with handle cut-out
    • Fold flat for easy storage and transport
    • All signs include same text and pictogram on both sides
    • All signs measure 20H x 12 in. W (51 x 30.5cm)

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit