Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Hazardous Waste Labels
Ensure the contents in containers are clearly identified and proper precautions are addressed with Container Identification Labels
Supplier: Accuform Signs MHZW11EVP
Specifications
Self-adhesive | |
Label | |
Hazardous Waste Label | |
Hazardous waste | |
6 x 6 in. | |
-20 to 176°F |
White, Green | |
Vinyl | |
Non Hazardous Waste Optional Information Shipper | |
25/Pk. | |
15.2 x 15.2 cm | |
-20°C to +80°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction