Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Information Centers
Post evacuation and assembly assignments, rescue and medical duties, security measures, and more in one convenient location
Supplier: Accuform Signs EMC205
Description
- .063 in. Aluminum backboard
- H x W: 20 x 15 in. (50.8 x 38.1cm)
- Printed with bold red stripes for high visibility
- Attached coated-wire basket with 1.5 in. dia. (3.81cm), three-ring binder
- Four corner mounting holes for easy installation
Specifications
Aluminum Backboard | |
Safety and Training Materials | |
Disaster Information Center |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction