Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Lockout Tags

Manufacturer:  ACCUFORM SIGNS MLT407LTP

 View more versions of this product

Catalog No. 19-804-840


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

5.87 x 3.37 in. (149 x 86mm)
Matte
5.87 in.
HS-Laminate
3.37 in.
Danger Do Not Operate
Identify crucial lockout or tagout points to allow maintenance to be performed safely
149mm
Lockout/Tagout Tag
86mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit