Accuform Signs NFPA Diamond Roll Labels

Manufacturer: ACCUFORM SIGNS LZN631PS2

 View more versions of this product

Catalog No. 19-121-454

  Check Availability
Add to Cart

    Specifications

    Format Label
    For Use With (Application) Hand written hazard codes and symbols
    Material Paper
    Adhesive Type Self-Adhesive
    Product Type NFPA Label
    Quantity 250 Roll(s)
    Color White
    Size (English) 2 x 2 in.
    Size (Metric) 5.08 x 5.08cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit