Learn More
Accuform Signs NFPA Diamond Roll Labels
Manufacturer:  ACCUFORM SIGNS LZN631EV2
 View more versions of this product
Catalog No. 19-121-455
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Self-adhesive
Hand written hazard codes and symbols
5.08 x 5.08cm
-40 to 121°C
Polyester
250 Roll(s)
White
2 x 2 in.
-40 to 250°F
Label
NFPA Label