Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs NFPA Placards

Designed for on-the-spot classification of potentially hazardous chemicals

Manufacturer:  ACCUFORM SIGNS ZFD100PA

 View more versions of this product

Catalog No. 19-121-452


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

10 x 10 in. (25.4 x 25.4cm)
10 in.
10 in.
Aluminum
-40 to 250°F
0.04 in.
10 in.
Excellent for indoor and outdoor usage, making it ideal for a wide variety of industrial strength applications
15 Numbers (3 sets of 0 to 4), 7 specific hazard symbols (Use No Water, Oxidizer, Simple Asphyxiant, Radiation Hazard, Corrosive, Acid and Alkali), NFPA wallet guide, blank placard
25.4cm
Each
-40 to 121°C
NFPA blank placard
25.4cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit