Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Pipe-Marking Tape
Solid color marking tapes increase pipe visibility and help to identify hazardous areas.
Supplier: Accuform Signs PTM613WT
Specifications
OSHA 1910.144 | |
White | |
1 Roll | |
-20 to 140°F | |
6 mil | |
32.92 m |
Self-adhesive | |
Vinyl | |
Pipe Marker | |
-28.88 to 60°C | |
108 ft. | |
1 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction