Accuform Signs Right To Know Center

Manufacturer: ACCUFORM SIGNS ZRS312

 View all options for this product

Catalog No. 19-805-307


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Category Safety and Training Materials
    For Use With (Application) Hold and store the MSDS binder
    Format Basket
    Product Type RTK Single Style Basket

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit