Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Roll-up Construction Signs

Signs safely control and regulate the flow of traffic through construction areas or other temporary work zones

Manufacturer:  ACCUFORM SIGNS FRC415MV

 View more versions of this product

Catalog No. 19-804-472


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Material: Mesh Vinyl
48 in.
Flagger Ahead
48 in.
Black, Orange
Each
Ideal for daytime use only
121.9cm
Roll-up
121.9cm
Vinyl
Diamond
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit