Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Safety Projection Sign: Emergency Shower

When you need identification, location, and direction, signs can be seen from various angles for extra visibility.

Manufacturer:  ACCUFORM SIGNS PSP710

 View more versions of this product

Catalog No. 17-998-101


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Green, White
8 in.
Each
10 in.
8 in.
Visual indicators to guide people to and mark the location of emergency showers or eyewash
Emergency Shower
90D
Projection sign
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit