Accuform Signs Safety Projection Sign: Spill Response Equipment

Locate equipment or direct your way out with multi-angle view

Manufacturer: ACCUFORM SIGNS PSP483

 View all options for this product

Catalog No. 17-998-093


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Certifications/Compliance OSHA 29 CFR 1910.145
    Color Green on White
    For Use With (Application) For identification, location and directional purposes
    Material Plastic
    Message Spill Response Equipment
    Quantity Each
    Size (English) 6 x 5 in.
    Style 90D
    Type Projection sign

    • High-impact plastic with permanently bonded image on the two panels
    • Protected by over-laminate film
    • 1.5 in. side flanged edges have pre-drilled mounting holes - includes screws and self-adhesive foam tape
    • 90D signs project at 90° from wall; measure 8 x 8 in.
    • 3D signs project at 45° from either side; measure 6 x 5 in.
    • Signs made of Lumi-Glow™ material glow in the dark

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit