Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Safety Signs, Bilingual: No Trespassing
to communicate possible hazards in the workplace or jobsite
$41.60 - $57.40
Specifications
Quantity | 1 Ea. |
---|---|
Max. Temperature (English) | 140°F |
Min. Temperature (English) | -20°F |
Temperature Range (Metric) | -28°C to 60°C |
Temperature Range (English) | -20°F to 140°F |
Catalog Number | Mfr. No. | Material | Displayed Text | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Displayed Text | Price | Quantity & Availability | ||||
19-804-992
|
Accuform Signs
SBMATR806VA |
Aluminum | Notice No Trespassing |
Each for $41.60
|
|
||||
19-804-998
|
Accuform Signs
SBMADM076XF |
Fiberglass | DANGER No Trespassing |
Each for $57.40
|
|
||||
Specifications
1 Ea. | |
-20°F | |
-20°F to 140°F |
140°F | |
-28°C to 60°C | |
Safety Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title