Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Safety Tags

Provide immediate identification of the hazard by using the appropriate message for your tag

Manufacturer:  ACCUFORM SIGNS MDT114PTP

 View more versions of this product

Catalog No. 17-998-139


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

OSHA 29 CFR 1910.145, ANSI Z535
0.37 in.
5.12 x 3.3 x 0.01 in. (13 x 8.38 x 0.02cm)
For indoor and outdoor conditions
0.02cm
13cm
15mil
8.38cm
Do Not Start
Black/Red on White
0.93cm
Matte
0.01 in.
5.12 in.
Standard back A
3.3 in.
RP Plastic
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit