Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Sprinlers Self-Stick Pipe Markers
Identify contents and mark the directional flow for pipes carrying sprinkler water.
Supplier: Accuform Signs RPK661SSD
Specifications
Pipe Marker | |
White, Red | |
Sprinkler | |
1 to 3 Years | |
-90 to 168°F | |
4 mil | |
6.35 to 15.24 cm |
Self-adhesive | |
Vinyl, Plastic | |
Clearly identify the pipe contents and flow direction | |
1 Ea. | |
-67.77 to 75.55°C | |
2.5 to 6 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction