Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ Lockout/Tagout Tags
Provide immediate identification of the hazard by using the appropriate message for your tag.
Supplier: Accuform™ MLT405CTP
Specifications
Tagout Tag | |
OSHA 29 CFR 1910.147 | |
Do Not Operate Equipment Lock Out | |
Ideal for indoor and short term outdoor conditions | |
147 mm | |
3.12 in. |
PF Cardstock | |
5.8 x 3.12 in. (147 x 79 mm) | |
Matte | |
5.8 in. | |
15 mil | |
79 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction