Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ Lockout/Tagout Tags
Provide immediate identification of the hazard by using the appropriate message for your tag.
Supplier: Accuform™ MLT409LTP
Specifications
Lockout/Tagout Tag | |
OSHA 29 CFR 1910.147 | |
Do Not Operate Equipment Locked Out By | |
Identify clearly and alert staff to cautions with durable safety tags that work effectively in extreme environments | |
142 mm | |
78 mm |
HS-Laminate | |
5.6 x 3 in. (142 x 78 mm) | |
Matte | |
5.6 in. | |
3 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction