Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter PROMO
Ideal for daily pH testing of your routine laboratory samples. The Accumet™ Basic AB315 Benchtop Laboratory pH/mV Meters provide the essential functions needed for common pH measurements, without complicated functions, at a budget-friendly price.
$445.00 - $500.00
Specifications
Measurement Modes | pH with mV and temperature, relative mV with mV and temperature |
---|---|
Calibration | pH, relative mV (RmV) and temperature |
Range (pH) | -2 to 18 |
Resolution (pH) | 0.01, 0.1 |
Accuracy (pH) | ±0.01 |
Catalog Number | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Type | Price | Quantity & Availability | ||||||
6A-B31-5AAC22
|
pH/mV Meter Standard Kit |
Each for $445.00
|
|
6AB315BAC22
|
pH/mV Meter Bio Kit |
Each for $500.00
|
|
Description
- Large, backlit display simultaneous shows pH, mV and temperature readings
- One to five point pH calibration with automatic buffer recognition of four pH buffer sets
- One point relative mV calibration (RmV) for ORP measurements
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon indicates measurement stability
- Electrode condition icon indicates electrode status
- 500 data set storage with time/date stamp
- Non-volatile meter memory preserves settings and data in event of power loss
- Meter-attached stand facilitates easy electrode movement into and out of samples
Specifications
pH with mV and temperature, relative mV with mV and temperature | |
-2 to 18 | |
±0.01 | |
±2000 | |
±0.2 or ±0.05%, whichever is greater | |
−5°C to +105°C | |
0.1°C (0.1°F) | |
LCD, 5 in. | |
Non-volatile | |
Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode | |
Calibration due alarm, selectable 1 to 60 hours or off | |
Communication port for computer or printer | |
IP54 | |
3 years | |
±2000.0 mV | |
Automatic or Manual | |
100/240 V |
pH, relative mV (RmV) and temperature | |
0.01, 0.1 | |
Selectable buffer sets: USA: 2.00, 4.01, 7.00, 10.01, 12.00; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1.00, 3.00, 6.00, 8.00, 10.00, 13.00 | |
1 mV, 0.1 RmV | |
1 to 5 (pH), 1 (relative mV), 1 (temperature) | |
23°F to 221°F | |
±0.3°C (±0.5°F) | |
Yes | |
500 | |
Printed or CSV Format | |
BNC, ATC | |
CE, TUV 3-1, FCC Class A | |
Included, 100-240 V Universal Power Adapter | |
AB315 | |
Indicator icons | |
1 | |
50/60 Hz |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title