Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

accumet™ pH 7.00 (Yellow) Buffer, Fisherbrand™

 View more versions of this product

Catalog No. 15-200-938


Add to Cart

Description

Description

Specifications

Specifications

pH Buffer, 5 gallon cubitainer
Accurate, reproducible pH calibrations
7
Buffer
Yes
pH 7.00 (Yellow) Buffer
pH Meter and Electrode
19 L
Cubitainer
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit