Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AccuPrime™ Pfx DNA Polymerase

AccuPrime™ Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis.

Supplier:  Invitrogen™ 12344024

Encompass_Preferred

 View more versions of this product

Catalog No. 12-344-024


Add to Cart

Includes: AccuPrime Pfx DNA Polymerase (100μL, at 2.5U/μL) is provided with 10X AccuPrime reaction mix (1mL) that contains dNTPs and a separate vial (1mL) of 50mM MgSO4. Reagents are provided for 200 amplification reactions of 50μL each.

Description

Description

AccuPrime™ Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcus species KOD with proofreading (3´→5´ exonuclease) activity. Platinum™ anti-Pfx DNA polymerase antibodies inhibit polymerase activity, providing hot-start capabilities for improved PCR specificity (see figure). Thermostable AccuPrime™ accessory proteins enhance specific primer-template hybridization during every cycle of PCR, increasing specificity, yield, and robustness over Platinum™ Pfx alone.

With AccuPrime™ Pfx DNA Polymerase you will:
• Achieve greater than 26 times higher fidelity than Taq DNA polymerase
• Amplify fragments up to 12 kb
• Minimize PCR optimization

Applications
Amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR. AccuPrime™ Pfx DNA Polymerase is especially effective where very high levels of fidelity are required.

Order Info

Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• AccuPrime Pfx DNA Polymerase (1 x 100 μL at 2.5 U/μL)
• 10X AccuPrime Pfx Reaction Mix (1 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -20°C.
Low
Standard
Wet or Dry Ice
26X
200 Reactions
Separate Components
100 μL
Primer-probe
AccuPrime Pfx DNA Polymerase
200 Reactions
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Blunt
12 kb or less
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.