Learn More
Pyramex™ Accurist™ Safety Eyewear
Click to view available options
Lens Tint:
Clear
Gray
Mirror, Silver
Silver
Frame Color:
Black
Black, Red
Description
Specifications
Certifications/Compliance | ANSI Z87.1,CSA Z94.3-07 |
Description | Full eye protection 10 curve lens |
Lens Tint | Clear |
Frame Color | Black |
Temple Style | Spatula with double injection rubber |
Lens Material | Polycarbonate |
Frame Material | Nylon |
Frame Type | Dual lens frame with side protection |