Learn More
Agilent Technologies ACCUSCRIPT HIGH 1ST STRAND
Supplier: Agilent Technologies 200820
AccuScript High Fidelity 1st Strand cDNA Synthesis Kit, 50 rxn, Contains: AccuScript RT, 10X RT Reaction buffer, RNase block ribonuclease inhibitor, 100mM dNTP mix, oligo(dT) and random primers, control RNA and control primers