Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ accuSkan™ FC Filter-Based Microplate Photometer
For research and routine ELISA applications
Description
A proven ELISA solution—accuSkan FC and the Fisher Scientific accuWash washer. The combination of the accuSkan FC microplate photometer and the accu-Wash™ microplate washer offers a convenient modular solution for routine and research ELISA applications.- Broad wavelength range of 340 to 850nm for a wide variety of applications
- Eight-position filter wheel with three standard pre-installed filters (405, 450 and 620nm)
- Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
- On-board linear shaking and optional incubation up to 50°C for temperature critical assays
- Superior usability with both internal software and dedicated PC software
- Data export capability via USB memory stick for easy data transfer
Specifications
Photodetector | |
PC Control, Internal Software | |
96 wells: 6 sec. (fast mode), 12 sec. (normal mode) | |
Linear shaking with 3 modes | |
accuSkan FC | |
8-position filter wheel; filters installed: 405nm, 450nm, and 620nm | |
96-well plate: 0 to 3 Abs (±2% at 405 nm) 384-well plate: 0 to 2.5 Abs (±2% at 405 nm) |
|
0.001 Abs | |
340 to 850 nm |
11.4 x 15.7 x 8.7 in. (29 x 40 x 22 cm) | |
Quartz Halogen Lamp | |
96-well Plates | |
±1% (0 to 3 Abs) or ±0.003 Abs, which ever is greater at 405 nm | |
100/240 V, 50/60 Hz | |
Robotic automation | |
CV ≤ 0.2% (0.3 to 3 Abs) at 405 nm | |
1 Year |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction