Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accuSkan™ FC Filter-Based Microplate Photometer - PROMO

For research and routine ELISA applications

 View more versions of this product

Catalog No. 14-377-575FBS


Add to cart

Description

Description

  • Broad wavelength range of 340 to 850nm for a wide variety of applications
  • Eight-position filter wheel with three standard pre-installed filters (405, 450 and 620nm)
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • On-board linear shaking and optional incubation up to 50°C for temperature critical assays
  • Superior usability with both internal software and dedicated PC software
  • Data export capability via USB memory stick for easy data transfer
Applications: Immunoassays (ELISA), protein and enzyme assays,cytotoxicity and proliferation assays, growth curves and endotoxins
Specifications

Specifications

accuSkan FC
96-well plates
Photodetector
340 to 850nm
0-3 Abs, ±2% at 405nm, 96-well plate 0-2.5 Abs, ±2% at 405nm, 384-well plate
±1% (0-3 Abs) or ±0.003 Abs, which ever is greater at 405nm
Linear shaking with three modes
Internal software or PC control
Robotic automation
1 Year
Quartz-halogen lamp
340 to 850nm
8-position filter wheel; filters installed: 405nm, 450nm, and 620nm
0.001 Abs
CV ≤0.2% (0.3-3 Abs) at 405nm
6 seconds, 96-well plate, fast mode; 12 seconds, 96-well plate, normal mode
100-240V 50/60Hz
11.4 x 15.7 x 8.7 in. (29 x 40 x 22cm)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit