Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Fisherbrand™ accuSkan™ GO UV/Vis Microplate Spectrophotometer PROMO

Measure both cuvettes and microplates, all according to your assay and throughput requirements. Fisherbrand™ accuSkan™ GO UV/Vis Microplate Spectrophotometer is a handy tool for photometric research applications.

 View more versions of this product

Catalog No. 14-377-579FBS



Description

Description

  • Select any measurement wavelength from low UV to visible with just a few clicks
  • Freely selectable wavelengths from 200 to 1000nm for the demands of various assays
  • Both microplate and cuvette reading for any throughput requirements
  • Fast plate measurements and a full sample spectrum in less than 10 seconds
  • Extensive self-diagnostics help ensure high quality data
  • Intuitive internal software for quick measurements
  • Easy and logical assay setup for demanding assays
Specifications

Specifications

One year
No
Monochromator
200 to 1000 nm with 1 nm steps
96-well plate: 0-2.5 Abs, 2% at 450 nm
1.0% + 0.003 Abs (0-2.0 Abs); 2% (2.0-2.5 Abs) at 450 nm
Ambient +2° to 45°C
6 seconds with 96-well plate; 10 seconds with 384-well plate (from A1 back to A1)
Stand-alone use: 4.5-inch color display and keypad; Dedicated PC Software
23.8 lb.
Max: <110w; Power Save: <2.5w
Robotic automation
96- and 384-well plates
Xenon flash
200 to 1000 nm with 1 nm steps
<2.5 nm
Up to 4 Abs
SD<0.003 Abs or CV<0.5% at 450 nm (Precision mode); SD<0.003 Abs or CV<1% at 450 nm (Fast mode)
Linear
10s from 200 to 1000 nm with 1 nm steps
10.8 kg
100-240 V, 50/60 Hz
Without cuvette
16.9 x 11.2 x 10.2 in. (43 x 28.5 x 26 cm)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit