Fisher Scientific™ accuSkan™ GO UV/Vis Microplate Spectrophotometer

Freedom for any photometric research application

 View more versions of this product

Catalog No. 14-377-579

  Check Availability
Add to Cart

    Specifications

    Bandwidth <2.5nm
    Excitation Wavelength Range 200 to 1000nm with 1nm steps
    Linearity 96-well plate: 0-2.5 Abs, 2% at 450nm
    Read-out Range Up to 4 Abs
    Accuracy 1.0% + 0.003 Abs (0-2.0 Abs); 2% (2.0-2.5 Abs) at 450nm
    Precision SD<0.003 Abs or CV<0.5% at 450nm (Precision mode); SD<0.003 Abs or CV<1% at 450nm (Fast mode)
    Incubator Temperature Ambient +2° to 45°C
    Shaking Linear
    Measurement Speed 6 seconds with 96-well plate; 10 seconds with 384-well plate (from A1 back to A1)
    Spectral Scanning Speed 10s from 200 to 1000nm with 1nm steps
    Interface Stand-alone use: 4.5-inch color display and keypad; Dedicated PC Software
    Weight (Metric) 10.8kg
    Weight (English) 23.8 lb.
    Electrical Requirements 100-240V, 50/60Hz
    Power Consumption Max: <110w; Power Save: <2.5w
    Includes accuSkan GO without cuvette
    Plate Type 96- and 384-well plates
    For Use With (Equipment) Robotic automation
    Light Source Xenon flash
    Wavelength Selection Monochromator
    Wavelength Range 200 to 1000nm with 1nm steps
    Warranty One year
    Cuvettes No
    Dimensions (L x W x H) 16.9 x 11.2 x 10.2 in. (43 x 28.5 x 26cm)

    The Fisher Scientific accuSkan GO microplate spectrophotometer is a handy tool for photometric research applications. Select any measurement wavelength from low UV to visible with just a few clicks. Measure both cuvettes and microplates, all according to your assay and throughput requirements.

    • Freely selectable wavelengths from 200 to 1000nm for the demands of various assays
    • Both microplate and cuvette reading for any throughput requirements
    • Fast plate measurements and a full sample spectrum in less than 10 seconds
    • Extensive self-diagnostics help ensure high quality data
    • Intuitive internal software for quick measurements
    • Easy and logical assay setup for demanding assays

    Applications: DNA or RNA quantitation and purity; Protein, enzyme and kinetic assays; Immunoassays; Cell proliferation and cytotoxicity assays; Apoptosis assays; Reporter gene assays; PCR assays

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit