Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accuWash™ and accuWash Versa Microplate Washers

Ease of use, convenience and versatility for washing microplates in ELISA applications

 View more versions of this product

Catalog No. 14-377-578


Add to cart

Includes: 2 x 2L wash bottle, 1 x 2L rinse bottle, 1 x 4L waste bottle, 2 x 8 wash head, aerosol cover

Description

Description

The Fisher Scientific accuWash and Fisher Scientific accuWash Versa are easy-to-use strip washers that offer convenience through a large color screen, intuitive graphical user interface, and a USB port. The accuWash is the basic model for washing 96-well plates only. It is intended for use when only a few similar assays are run routinely. The accuWash Versa can also wash cells and 384-well plates, offering enhanced flexibility needed for research use.

Key Features
  • Easy protocol setup with the large color screen and the visual internal software
  • A USB port for transferring protocols between several washers, and for easy updating of the internal software
  • Sweep mode provides an extremely low residual volume in the well, resulting in excellent washing performance and reliable assay results
Easy and convenient to use
  • Large color screen shows everything needed for normal use on one display
  • Internal software is logical and intuitive, and allows protocols to be developed fast, making it easy to get started with the softwaredash (em)requires minimal training
Reliable and secure performance
  • Unpressurized bottles are safe and secure to use
  • Automatic rinse and prime features prevent clogging of liquid transfer system
  • Liquid level sensors provide security for both wash and waste bottles
  • Low residual volumes of washers provide optimal washing performance, helping deliver reliable assay results
Added versatility
  • Washes cells, 96- and 384-well plates
  • Compatable with automation systems
Applications: Immunoassays (ELISA), Cell washing (accuWash Versa)

Certifications

CE mark

Warranty and Services

1 year

Specifications

Specifications

96- and 384-Well Plates
1x8, 1x12, 2x8, 2x12, and 1x16 wash heads; 2x8 cell wash head
less than 2μL
Yes
9kg
Automation systems
100-240V
Immunoassays (ELISA); Cell Washing
accuWash Versa
Up to 3
50 to 1,000μL (96); 20 to 300μL (384)
50 to 400μL (96); 20 to 120μL (384)
LCD Color Display, 4.3 in.
20 lb.
2x2L wash bottle, 2 x 1L rinse bottle, 1x4L waste bottle, 2x8 wash head, aerosol cover
USB, RS-232-C
100-240V, 50/60Hz
15.2 x 13.6 x 9.4 in. (38.5 x 34.5 x 24cm)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit