Fisher Scientific™ accuWash™ and accuWash Versa Microplate Washers

Ease of use, convenience and versatility for washing microplates in ELISA applications

 View more versions of this product

Catalog No. 14-377-577

  Check Availability
Add to Cart

Includes: 1x2L wash bottle, 1x2L waste bottle, 1x8 wash head, aerosol cover

    Specifications

    Wash/Waste Bottle Capacity 1
    Plate Type 96-Well Plates
    Cell Washing No
    Display LCD Color Display, 4.3 in.
    Weight (Metric) 9kg
    Weight (English) 20 lb.
    Includes 1x2L wash bottle, 1x2L waste bottle, 1x8 wash head, aerosol cover
    Voltage 100-240V
    Wash Heads 1 x 8; 1 x 12
    Connections USB
    For Use With (Application) ELISA-Based Assays; Cell Washing
    Wash Volume 50 to 1,000μL
    Electrical Requirements 100-240V, 50/60Hz
    Description accuWash
    Dimensions (L x W x H) 15.2 x 13.6 x 9.4 in. (38.5 x 34.5 x 24cm)
    Residual Volume less than 2μL
    Dispense Volume 50 to 400μL (96)

    The Fisher Scientific accuWash and Fisher Scientific accuWash Versa are easy-to-use strip washers that offer convenience through a large color screen, intuitive graphical user interface, and a USB port. The accuWash is the basic model for washing 96-well plates only. It is intended for use when only a few similar assays are run routinely. The accuWash Versa can also wash cells and 384-well plates, offering enhanced flexibility needed for research use.

    Certifications

    CE mark

    Warranty

    1 year

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit