Learn More
Aviva Systems Biology Corp ACD Antibody - C-terminal region (ARP58254_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58254_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ACD. Alternate names: PIP1, PTOP, TINT1, TPP1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62271